Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cap-D3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001169416.2 | NP_001162887.1 | 2 - 3 | 1036 | 58 | |
NM_135091.4 | NP_608935.2 | 2 - 3 | 1036 | 58 |
Set Membership: |
Amplicon length less than or equal to 70 Drosophila melanogaster |