Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: AGO2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001274953.1 | NP_001261882.1 | 5 - 6 | 3388 | 67 | |
NM_140518.3 | NP_648775.1 | 7 - 8 | 3654 | 67 | ||
NM_168626.3 | NP_730054.1 | 6 - 7 | 3721 | 67 | ||
GenBank mRNA | BT099682.1 | - | 6 - 7 | 3659 | 67 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Drosophila melanogaster |