Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: comm2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_140544.2 | NP_648801.2 | 1 - 2 | 652 | 71 | |
GenBank mRNA | AY113389.1 | - | 1 - 2 | 651 | 71 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Recommended Assay Drosophila melanogaster |