Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Exn | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001104160.3 | NP_001097630.2 | 8 - 9 | 6581 | 67 | |
NM_001202194.1 | NP_001189123.1 | 8 - 9 | 6481 | 67 | ||
NM_140685.5 | NP_648942.1 | 7 - 8 | 3093 | 67 | ||
NM_168705.2 | NP_730228.1 | 7 - 8 | 3100 | 67 | ||
NM_168706.2 | NP_730229.1 | 7 - 8 | 3069 | 67 | ||
GenBank mRNA | AF145619.1 | - | 6 - 7 | 3026 | 67 | |
BT132960.1 | - | 8 - 9 | 6481 | 67 | ||
EF439708.1 | - | 7 - 8 | 3401 | 67 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Recommended Assay Drosophila melanogaster |