Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Vps60 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001275065.1 | NP_001261994.1 | 3 - 4 | 633 | 76 | |
NM_140740.4 | NP_648997.1 | 3 - 4 | 513 | 76 | ||
GenBank mRNA | AY071206.1 | - | 3 - 4 | 506 | 76 |
Set Membership: |
Amplicon length between 71 and 85 Drosophila melanogaster |