Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cyp12c1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001300180.1 | NP_001287109.1 | 1 - 2 | 452 | 90 | |
NM_140795.3 | NP_649052.1 | 1 - 2 | 452 | 90 | ||
GenBank mRNA | AY058426.1 | - | 1 - 2 | 437 | 90 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Recommended Assay Drosophila melanogaster |