Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: wnd | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_140880.3 | NP_649137.3 | 2 - 3 | 455 | 66 | |
NM_176362.2 | NP_788540.1 | 2 - 3 | 378 | 66 | ||
NM_176363.3 | NP_788541.1 | 2 - 3 | 583 | 66 | ||
GenBank mRNA | AY094787.1 | - | 2 - 3 | 405 | 66 |
Set Membership: |
Amplicon length less than or equal to 70 Drosophila melanogaster |