Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: tey | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001144509.3 | NP_001137981.1 | 6 - 7 | 1903 | 72 | |
NM_001275130.2 | NP_001262059.1 | 6 - 7 | 1873 | 72 | ||
NM_001275131.1 | NP_001262060.1 | 6 - 7 | 1718 | 72 | ||
NM_140883.2 | NP_649140.1 | 6 - 7 | 1903 | 72 |
Set Membership: |
Amplicon length between 71 and 85 Drosophila melanogaster |