Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: tey | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_140883.2 | NP_649140.1 | 8 - 9 | 2421 | 61 | |
GenBank mRNA | BT133456.1 | - | 8 - 9 | 2420 | 61 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Drosophila melanogaster |