Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rbbp5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_140952.3 | NP_649209.1 | 6 - 7 | 1413 | 79 | |
GenBank mRNA | AY071120.1 | - | 6 - 7 | 1410 | 79 | |
BT032759.1 | - | 6 - 7 | 1418 | 79 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Drosophila melanogaster |