Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: bru1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001169492.2 | NP_001162963.1 | 8 - 9 | 1882 | 60 | |
NM_001273474.1 | NP_001260403.1 | 9 - 10 | 2520 | 60 | ||
NM_001273475.1 | NP_001260404.1 | 8 - 9 | 1754 | 60 | ||
NM_001273476.1 | NP_001260405.1 | 8 - 9 | 1607 | 60 | ||
NM_001273477.1 | NP_001260406.1 | 8 - 9 | 1912 | 60 | ||
NM_001273478.1 | NP_001260407.1 | 8 - 9 | 1883 | 60 | ||
NM_165002.2 | NP_723737.1 | 9 - 10 | 2667 | 60 | ||
NM_165003.3 | NP_723738.2 | 10 - 11 | 1930 | 60 | ||
NM_165004.3 | NP_723739.1 | 8 - 9 | 1882 | 60 | ||
NM_165005.3 | NP_723740.1 | 6 - 7 | 1866 | 60 | ||
GenBank mRNA | AY051758.1 | - | 8 - 9 | 1862 | 60 | |
AY071650.1 | - | 6 - 7 | 1864 | 60 | ||
BT133212.1 | - | 9 - 10 | 2646 | 60 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Recommended Assay Drosophila melanogaster |