Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: bru2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001042891.3 | NP_001036356.1 | 6 - 7 | 1949 | 62 | |
NM_001259069.2 | NP_001245998.1 | 5 - 6 | 1773 | 62 | ||
NM_001259070.1 | NP_001245999.1 | 3 - 4 | 869 | 62 | ||
NM_001259071.2 | NP_001246000.1 | 6 - 7 | 1949 | 62 | ||
NM_001259072.1 | NP_001246001.1 | 2 - 3 | 431 | 62 | ||
NM_001259073.1 | NP_001246002.1 | 3 - 4 | 960 | 62 | ||
NM_001273481.1 | NP_001260410.1 | 6 - 7 | 2232 | 62 | ||
NM_001273482.1 | NP_001260411.1 | 6 - 7 | 1856 | 62 | ||
NM_135715.4 | NP_609559.1 | 6 - 7 | 1856 | 62 | ||
NM_165007.3 | NP_723742.2 | 6 - 7 | 1856 | 62 | ||
NM_176025.3 | NP_788039.1 | 6 - 7 | 1949 | 62 | ||
GenBank mRNA | AY069464.1 | - | 6 - 7 | 1823 | 62 | |
BT081408.1 | - | 3 - 4 | 796 | 62 |
Set Membership: |
Amplicon length less than or equal to 70 Recommended Assay Drosophila melanogaster |