Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pect | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_135769.3 | NP_609613.2 | 3 - 4 | 825 | 75 | |
NM_165028.2 | NP_723789.2 | 3 - 4 | 825 | 75 | ||
NM_165029.2 | NP_723790.2 | 3 - 4 | 825 | 75 | ||
NM_165030.2 | NP_723791.2 | 3 - 4 | 825 | 75 | ||
GenBank mRNA | BT003190.1 | - | 3 - 4 | 819 | 75 | |
BT125823.1 | - | 3 - 4 | 536 | 75 |
Set Membership: |
Amplicon length between 71 and 85 Drosophila melanogaster |