Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: kuz | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_057839.5 | NP_477187.1 | 12 - 13 | 4660 | 101 | |
NM_165047.3 | NP_723825.1 | 12 - 13 | 4533 | 101 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Drosophila melanogaster |