Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ck | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001299020.1 | NP_001285949.1 | 10 - 11 | 6468 | 139 | |
NM_001299021.1 | NP_001285950.1 | 10 - 11 | 6631 | 139 | ||
NM_078847.4 | NP_523571.1 | 10 - 11 | 6463 | 139 | ||
NM_165099.3 | NP_723895.1 | 10 - 11 | 6468 | 139 | ||
GenBank mRNA | BT132752.1 | - | 10 - 11 | 6415 | 139 |
Set Membership: |
Amplicon length greater than or equal to 101 Drosophila melanogaster |