Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gli | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_057254.4 | NP_476602.1 | 6 - 7 | 2459 | 85 | |
NM_165127.2 | NP_723928.1 | 7 - 8 | 2550 | 85 | ||
NM_165128.2 | NP_723929.1 | 6 - 7 | 2339 | 85 | ||
NM_165129.2 | NP_723930.1 | 6 - 7 | 2371 | 85 | ||
NM_165130.2 | NP_723931.2 | 6 - 7 | 2832 | 85 | ||
GenBank mRNA | BT133203.1 | - | 6 - 7 | 2456 | 85 |
Set Membership: |
Amplicon length between 71 and 85 Drosophila melanogaster |