Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hcf | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001258491.3 | NP_001245420.1 | 11 - 12 | 4207 | 103 | |
NM_079882.4 | NP_524621.2 | 10 - 11 | 4181 | 103 | ||
NM_166756.3 | NP_726566.1 | 10 - 11 | 3877 | 103 | ||
NM_166757.3 | NP_726567.2 | 10 - 11 | 4025 | 103 | ||
NM_205873.3 | NP_995595.1 | 10 - 11 | 3877 | 103 | ||
GenBank mRNA | AJ320236.1 | - | 9 - 10 | 3834 | 103 | |
BT015268.1 | - | 10 - 11 | 4142 | 103 |
Set Membership: |
Amplicon length greater than or equal to 101 Drosophila melanogaster |