Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Eaat2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001169373.2 | NP_001162844.1 | 1 - 2 | 982 | 66 | |
NM_001169374.2 | NP_001162845.1 | 3 - 4 | 753 | 66 | ||
NM_058079.3 | NP_477427.1 | 3 - 4 | 641 | 66 |
Set Membership: |
Amplicon length less than or equal to 70 Drosophila melanogaster |