Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: bchs | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_135124.2 | NP_608968.2 | 10 - 11 | 8704 | 70 |
Set Membership: |
Amplicon length less than or equal to 70 Drosophila melanogaster |