Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: retm | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001273203.2 | NP_001260132.1 | 1 - 2 | 400 | 70 | |
NM_001298743.1 | NP_001285672.1 | 1 - 2 | 400 | 70 | ||
NM_135184.4 | NP_609028.2 | 1 - 2 | 400 | 70 | ||
GenBank mRNA | BT044154.1 | - | 1 - 2 | 335 | 70 |
Set Membership: |
Amplicon length less than or equal to 70 Drosophila melanogaster |