Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: COX5B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001298749.1 | NP_001285678.1 | 1 - 2 | 189 | 67 | |
NM_135202.2 | NP_609046.1 | 1 - 2 | 189 | 67 | ||
GenBank mRNA | AY070965.1 | - | 1 - 2 | 187 | 67 |
Set Membership: |
Amplicon length less than or equal to 70 Drosophila melanogaster |