Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GATAd | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001273397.1 | NP_001260326.1 | 2 - 3 | 1225 | 94 | |
NM_001273398.1 | NP_001260327.1 | 2 - 3 | 1045 | 94 | ||
NM_135539.3 | NP_609383.2 | 2 - 3 | 1045 | 94 | ||
GenBank mRNA | BT133312.1 | - | 2 - 3 | 1037 | 94 |
Set Membership: |
Amplicon length between 86 and 100 Drosophila melanogaster |