Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RluA-2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001201853.2 | NP_001188782.2 | 7 - 8 | 1293 | 90 | |
NM_001201854.2 | NP_001188783.1 | 6 - 7 | 1426 | 90 | ||
NM_001273423.1 | NP_001260352.1 | 6 - 7 | 1401 | 90 | ||
NM_135567.4 | NP_609411.1 | 6 - 7 | 1426 | 90 | ||
GenBank mRNA | AY058686.1 | - | 6 - 7 | 1378 | 90 |
Set Membership: |
Amplicon length between 86 and 100 Drosophila melanogaster |