Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: spir | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001169549.1 | NP_001163020.1 | 3 - 4 | 1386 | 68 | |
NM_001259172.2 | NP_001246101.1 | 3 - 4 | 1386 | 68 | ||
NM_001259173.1 | NP_001246102.1 | 3 - 4 | 1386 | 68 | ||
NM_001259174.1 | NP_001246103.1 | 3 - 4 | 1386 | 68 | ||
NM_001299170.1 | NP_001286099.1 | 3 - 4 | 1386 | 68 | ||
NM_001299171.1 | NP_001286100.1 | 3 - 4 | 1386 | 68 | ||
NM_080115.3 | NP_524854.2 | 3 - 4 | 1386 | 68 | ||
NM_165323.3 | NP_724254.1 | 3 - 4 | 1386 | 68 | ||
NM_165324.2 | NP_724255.1 | 3 - 4 | 1386 | 68 | ||
GenBank mRNA | AJ238876.1 | - | 3 - 4 | 1033 | 68 | |
AY075586.1 | - | 3 - 4 | 1372 | 68 | ||
BT122081.1 | - | 3 - 4 | 1065 | 68 |
Set Membership: |
Amplicon length less than or equal to 70 Drosophila melanogaster |