Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: spir | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001169549.1 | NP_001163020.1 | 7 - 8 | 1825 | 68 | |
NM_001259172.2 | NP_001246101.1 | 7 - 8 | 1822 | 68 | ||
NM_001259173.1 | NP_001246102.1 | 7 - 8 | 1828 | 68 | ||
NM_001259174.1 | NP_001246103.1 | 7 - 8 | 1822 | 68 | ||
NM_001299170.1 | NP_001286099.1 | 8 - 9 | 1852 | 68 | ||
NM_001299171.1 | NP_001286100.1 | 7 - 8 | 1828 | 68 | ||
NM_080115.3 | NP_524854.2 | 7 - 8 | 1828 | 68 | ||
NM_165323.3 | NP_724254.1 | 7 - 8 | 1828 | 68 | ||
NM_165324.2 | NP_724255.1 | 7 - 8 | 1825 | 68 | ||
GenBank mRNA | AJ238876.1 | - | 7 - 8 | 1475 | 68 | |
BT122081.1 | - | 7 - 8 | 1504 | 68 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Drosophila melanogaster |