Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: IKKepsilon | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_136204.3 | NP_610048.2 | 3 - 4 | 1294 | 87 | |
NM_165337.2 | NP_724278.1 | 2 - 3 | 1204 | 87 | ||
GenBank mRNA | BT046140.2 | - | 2 - 3 | 1178 | 87 |
Set Membership: |
Amplicon length between 86 and 100 Recommended Assay Drosophila melanogaster |