Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: dia | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001259184.1 | NP_001246113.1 | 3 - 4 | 2422 | 59 | |
NM_001273711.1 | NP_001260640.1 | 3 - 4 | 2784 | 59 | ||
NM_057633.5 | NP_476981.1 | 3 - 4 | 2784 | 59 | ||
NM_165341.3 | NP_724285.1 | 3 - 4 | 2506 | 59 | ||
GenBank mRNA | BT021398.1 | - | 3 - 4 | 2762 | 59 |
Set Membership: |
Amplicon length less than or equal to 70 Drosophila melanogaster |