Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: tadr | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_136238.3 | NP_610082.2 | 3 - 4 | 652 | 59 | |
NM_165354.2 | NP_724308.2 | 3 - 4 | 652 | 59 |
Set Membership: |
Amplicon length less than or equal to 70 Recommended Assay Drosophila melanogaster |