Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: vlc | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001169585.3 | NP_001163056.1 | 2 - 3 | 354 | 78 | |
NM_001169586.1 | NP_001163057.1 | 1 - 2 | 269 | 78 | ||
NM_001259213.1 | NP_001246142.1 | 2 - 3 | 215 | 78 | ||
NM_001259214.2 | NP_001246143.1 | 2 - 3 | 97 | 78 | ||
NM_080129.5 | NP_524868.1 | 2 - 3 | 259 | 78 | ||
NM_165442.5 | NP_724425.1 | 2 - 3 | 182 | 78 | ||
NM_165443.4 | NP_724426.3 | 2 - 3 | 153 | 78 | ||
NM_165444.5 | NP_724427.1 | 2 - 3 | 182 | 78 | ||
GenBank mRNA | AF275628.1 | - | 1 - 2 | 88 | 78 | |
AY061484.1 | - | 2 - 3 | 238 | 78 | ||
BT100113.1 | - | 2 - 3 | 182 | 78 |
Set Membership: |
Amplicon length between 71 and 85 Drosophila melanogaster |