Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: EcR | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_165461.3 | NP_724456.1 | 3 - 4 | 981 | 78 | |
NM_165462.2 | NP_724457.1 | 4 - 5 | 1060 | 78 | ||
NM_165463.2 | NP_724458.1 | 4 - 5 | 1106 | 78 |
Set Membership: |
Amplicon length between 71 and 85 Drosophila melanogaster |