Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: bin3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001273795.1 | NP_001260724.1 | 4 - 5 | 2673 | 64 | |
NM_001316407.1 | NP_001303336.1 | 4 - 5 | 2778 | 64 | ||
NM_078902.3 | NP_523626.2 | 4 - 5 | 2715 | 64 | ||
NM_165468.2 | NP_724468.1 | 4 - 5 | 2768 | 64 | ||
GenBank mRNA | AY052142.1 | - | 4 - 5 | 2745 | 64 |
Set Membership: |
Amplicon length less than or equal to 70 Drosophila melanogaster |