Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: wech | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001144142.3 | NP_001137614.1 | 3 - 4 | 2556 | 72 | |
NM_001144143.2 | NP_001137615.1 | 4 - 5 | 2632 | 72 | ||
NM_080033.4 | NP_524772.2 | 5 - 6 | 2811 | 72 | ||
NM_165533.3 | NP_724567.1 | 4 - 5 | 2886 | 72 | ||
NM_165534.3 | NP_724568.1 | 4 - 5 | 2886 | 72 | ||
GenBank mRNA | BT010087.1 | - | 4 - 5 | 2846 | 72 | |
BT030403.1 | - | 5 - 6 | 2826 | 72 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Drosophila melanogaster |