Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: nito | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001299245.1 | NP_001286174.1 | 2 - 3 | 770 | 89 | |
NM_136495.3 | NP_610339.1 | 2 - 3 | 770 | 89 | ||
NM_165579.2 | NP_724633.1 | 2 - 3 | 770 | 89 | ||
GenBank mRNA | AF145664.1 | - | 2 - 3 | 747 | 89 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Recommended Assay Drosophila melanogaster |