Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: rgr | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_165611.3 | NP_724690.1 | 1 - 2 | 818 | 58 | |
GenBank mRNA | AF353512.1 | - | 1 - 2 | 766 | 58 | |
BT150319.1 | - | 1 - 2 | 789 | 58 |
Set Membership: |
Amplicon length less than or equal to 70 Drosophila melanogaster |