Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: trpl | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_057547.5 | NP_476895.1 | 9 - 10 | 2105 | 78 | |
NM_165694.3 | NP_724822.1 | 9 - 10 | 2230 | 78 | ||
NM_165695.2 | NP_724823.2 | 9 - 10 | 2092 | 78 | ||
GenBank mRNA | BT099607.1 | - | 9 - 10 | 2076 | 78 |
Set Membership: |
Amplicon length between 71 and 85 Drosophila melanogaster |