Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Oatp33Ea | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001273488.1 | NP_001260417.1 | 4 - 5 | 1601 | 75 | |
NM_135724.4 | NP_609568.3 | 4 - 5 | 1589 | 75 | ||
GenBank mRNA | BT125052.1 | - | 4 - 5 | 1538 | 75 |
Set Membership: |
Amplicon length between 71 and 85 Drosophila melanogaster |