Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Oatp33Eb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_135726.3 | NP_609570.1 | 1 - 2 | 246 | 92 | |
GenBank mRNA | AY071008.1 | - | 1 - 2 | 244 | 92 |
Set Membership: |
Amplicon length between 86 and 100 Drosophila melanogaster |