Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nep1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001297996.1 | NP_001284925.1 | 7 - 8 | 2593 | 71 | |
NM_001297997.1 | NP_001284926.1 | 8 - 9 | 3062 | 71 | ||
NM_078501.3 | NP_511056.2 | 7 - 8 | 2404 | 71 | ||
NM_167059.2 | NP_727065.1 | 7 - 8 | 2425 | 71 | ||
GenBank mRNA | BT003622.1 | - | 7 - 8 | 2425 | 71 |
Set Membership: |
Amplicon length between 71 and 85 Drosophila melanogaster |