Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hk | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001258680.2 | NP_001245609.1 | 12 - 13 | 2747 | 65 | |
NM_001258681.2 | NP_001245610.1 | 9 - 10 | 2489 | 65 | ||
NM_001258682.2 | NP_001245611.1 | 11 - 12 | 2616 | 65 | ||
NM_001258683.2 | NP_001245612.1 | 11 - 12 | 2237 | 65 | ||
NM_001272470.2 | NP_001259399.1 | 6 - 7 | 2252 | 65 | ||
NM_001272471.2 | NP_001259400.1 | 6 - 7 | 2249 | 65 | ||
NM_001272472.2 | NP_001259401.1 | 6 - 7 | 2249 | 65 | ||
NM_078549.4 | NP_511104.3 | 6 - 7 | 2249 | 65 | ||
NM_167222.3 | NP_727405.1 | 11 - 12 | 2234 | 65 | ||
GenBank mRNA | AY060359.1 | - | 6 - 7 | 1378 | 65 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Drosophila melanogaster Inventoried |