Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Or10a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_078567.5 | NP_511122.1 | 2 - 3 | 747 | 130 |
Gene Symbol: Gr10a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_167283.3 | NP_727523.1 | 2 - 3 | 747 | 130 |
Set Membership: |
Amplicon length greater than or equal to 101 G-protein coupled receptor Drosophila melanogaster |