Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ten-a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001144717.4 | NP_001138189.1 | 12 - 13 | 4565 | 68 | |
NM_001144718.3 | NP_001138190.1 | 13 - 14 | 5270 | 68 | ||
NM_001169258.2 | NP_001162729.1 | 7 - 8 | 2499 | 68 | ||
NM_001169259.3 | NP_001162730.2 | 13 - 14 | 4675 | 68 | ||
NM_001169260.3 | NP_001162731.1 | 14 - 15 | 5541 | 68 | ||
NM_001169261.3 | NP_001162732.2 | 12 - 13 | 4565 | 68 | ||
NM_001258711.2 | NP_001245640.1 | 10 - 11 | 3514 | 68 | ||
NM_001258712.2 | NP_001245641.1 | 10 - 11 | 3520 | 68 | ||
NM_001258713.2 | NP_001245642.1 | 10 - 11 | 3532 | 68 | ||
NM_001272554.2 | NP_001259483.1 | 10 - 11 | 3414 | 68 | ||
NM_078582.4 | NP_511137.3 | 8 - 9 | 2700 | 68 |
Set Membership: |
Amplicon length less than or equal to 70 Drosophila melanogaster |