Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Flo2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001169287.2 | NP_001162758.1 | 4 - 5 | 1226 | 106 | |
NM_001169288.1 | NP_001162759.1 | 3 - 4 | 893 | 106 | ||
NM_001272623.2 | NP_001259552.1 | 6 - 7 | 1274 | 106 | ||
NM_001272624.2 | NP_001259553.1 | 7 - 8 | 1313 | 106 | ||
NM_001272625.2 | NP_001259554.1 | 6 - 7 | 1244 | 106 | ||
NM_001272626.2 | NP_001259555.1 | 7 - 8 | 1624 | 106 | ||
NM_078602.4 | NP_511157.2 | 6 - 7 | 1244 | 106 | ||
NM_167415.3 | NP_727797.1 | 7 - 8 | 1283 | 106 | ||
NM_167416.3 | NP_727798.1 | 7 - 8 | 1395 | 106 | ||
NM_167417.3 | NP_727799.1 | 6 - 7 | 1356 | 106 | ||
NM_167419.2 | NP_727812.2 | 3 - 4 | 1636 | 106 | ||
NM_167421.2 | NP_727814.1 | 3 - 4 | 1206 | 106 | ||
GenBank mRNA | BT009962.1 | - | 6 - 7 | 1242 | 106 | |
BT099913.1 | - | 3 - 4 | 1206 | 106 | ||
BT126133.1 | - | 7 - 8 | 1262 | 106 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Recommended Assay Drosophila melanogaster |