Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: eag | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001042810.3 | NP_001036275.1 | 4 - 5 | 1165 | 62 | |
NM_001201662.2 | NP_001188591.1 | 4 - 5 | 1165 | 62 | ||
NM_001272627.1 | NP_001259556.1 | 4 - 5 | 1165 | 62 | ||
NM_001272628.1 | NP_001259557.1 | 4 - 5 | 1165 | 62 | ||
NM_001272629.1 | NP_001259558.1 | 4 - 5 | 1165 | 62 | ||
NM_078603.4 | NP_511158.2 | 4 - 5 | 1165 | 62 | ||
GenBank mRNA | BT150281.1 | - | 2 - 3 | 615 | 62 |
Set Membership: |
Amplicon length less than or equal to 70 Drosophila melanogaster |