Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pkcdelta | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001144719.2 | NP_001138191.1 | 14 - 15 | 2399 | 69 | |
NM_078616.4 | NP_511171.3 | 14 - 15 | 2575 | 69 | ||
NM_132569.3 | NP_572797.3 | 7 - 8 | 5644 | 69 | ||
GenBank mRNA | BT021275.1 | - | 13 - 14 | 2331 | 69 |
Set Membership: |
Amplicon length less than or equal to 70 Drosophila melanogaster |