Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pkcdelta | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001144719.2 | NP_001138191.1 | 15 - 16 | 2537 | 59 | |
NM_078616.4 | NP_511171.3 | 15 - 16 | 2713 | 59 | ||
NM_132569.3 | NP_572797.3 | 8 - 9 | 5782 | 59 | ||
GenBank mRNA | BT021275.1 | - | 14 - 15 | 2469 | 59 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Drosophila melanogaster |