Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
CDMO and Clinical Trial Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Instrument Management
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
Home
›
TaqMan
®
Gene Expression Assays
›
Search Tool
›
Search Results
› Dm01813746_g1
See other PARA GE Assays ›
Gene Symbol:
para
Gene Name:
paralytic
Gene Aliases:
CG9907, DmNa[[V]], DmNa[[v]], DmNa[[v]]1, DmNav, DmNav1, Dmel\CG9907, Dmel_CG9907, Nav1, Ocd, Para, Shu, Shudderer, VSSC, bas, bss, l(1)14Da, l(1)ESHS48, lincRNA.S9469, olfD, par, sbl, sbl-1
Chromosome Location:
Chr.X: 16455027 - 16533096 on Build Release_6_plus_ISO1_MT
Species:
Drosophila melanogaster
Species Specific ID (Flybase ID):
FBgn0285944
Assay ID
Dm01813746_g1
Size
FAM-MGB | XS: 75 rxns
FAM-MGB | S: 360 rxns
FAM-MGB | M: 750 rxns
FAM-MGB | L: 2900 rxns
VIC-MGB_PL | S: 360 rxns
VIC-MGB_PL | M: 750 rxns
VIC-MGB_PL | L: 2900 rxns
VIC-MGB | S: 360 rxns
VIC-MGB | M: 750 rxns
VIC-MGB | L: 2900 rxns
Availability
Made To Order
Catalog #
4351372
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
Assay Details
More Information
Genomic Map
LOADING...
×
Back To Top
Assay Details
Gene Symbol: para
Interrogated Sequence
Translated Protein
Exon Boundary
Assay Location
IMAGE Clone ID
Amplicon Length
RefSeq
NM_001042815.2
NP_001036280.1
8 - 9
1034
58
NM_001042816.2
NP_001036281.1
8 - 9
1034
58
NM_001144733.2
NP_001138205.1
7 - 8
1001
58
NM_001201671.1
NP_001188600.1
8 - 9
1034
58
NM_001201672.1
NP_001188601.1
8 - 9
1034
58
NM_001201673.1
NP_001188602.1
8 - 9
1034
58
NM_001201674.1
NP_001188603.1
7 - 8
1001
58
NM_001201675.1
NP_001188604.1
8 - 9
1034
58
NM_001201676.1
NP_001188605.1
8 - 9
1034
58
NM_001201677.1
NP_001188606.1
8 - 9
1034
58
NM_001201678.1
NP_001188607.1
8 - 9
1034
58
NM_001201679.1
NP_001188608.1
8 - 9
1034
58
NM_001201680.1
NP_001188609.1
8 - 9
1034
58
NM_001201681.1
NP_001188610.1
8 - 9
1034
58
NM_001201682.1
NP_001188611.1
8 - 9
1034
58
NM_001201683.1
NP_001188612.1
8 - 9
1034
58
NM_001201684.1
NP_001188613.1
8 - 9
1034
58
NM_001201685.1
NP_001188614.1
8 - 9
1034
58
NM_001201686.1
NP_001188615.1
8 - 9
1034
58
NM_001201687.1
NP_001188616.1
8 - 9
1034
58
NM_001201688.1
NP_001188617.1
8 - 9
1034
58
NM_001201689.1
NP_001188618.1
8 - 9
1034
58
NM_001201690.1
NP_001188619.1
8 - 9
1034
58
NM_001201691.1
NP_001188620.1
8 - 9
1034
58
NM_001201692.1
NP_001188621.1
8 - 9
1034
58
NM_001201693.1
NP_001188622.1
8 - 9
1034
58
NM_001201694.1
NP_001188623.1
8 - 9
1034
58
NM_001201695.2
NP_001188624.1
8 - 9
1034
58
NM_001201696.1
NP_001188625.1
7 - 8
1001
58
NM_001201697.1
NP_001188626.1
8 - 9
1034
58
NM_001201698.1
NP_001188627.1
7 - 8
1001
58
NM_001201699.1
NP_001188628.1
7 - 8
1001
58
NM_001201700.1
NP_001188629.1
7 - 8
1001
58
NM_001201701.1
NP_001188630.1
7 - 8
1001
58
NM_001201702.1
NP_001188631.1
7 - 8
1001
58
NM_001201703.3
NP_001188632.1
8 - 9
1524
58
NM_001201704.1
NP_001188633.1
7 - 8
1001
58
NM_001201705.1
NP_001188634.1
7 - 8
1001
58
NM_001201706.1
NP_001188635.1
8 - 9
1034
58
NM_001201707.1
NP_001188636.1
8 - 9
1034
58
NM_001201708.1
NP_001188637.1
8 - 9
1034
58
NM_001201709.1
NP_001188638.1
8 - 9
1034
58
NM_001201711.1
NP_001188640.1
6 - 7
872
58
NM_001201712.1
NP_001188641.1
6 - 7
872
58
NM_001201713.1
NP_001188642.1
6 - 7
872
58
NM_001201714.1
NP_001188643.1
6 - 7
872
58
NM_001201715.1
NP_001188644.1
7 - 8
1001
58
NM_001201716.1
NP_001188645.1
7 - 8
1001
58
NM_001201717.1
NP_001188646.1
7 - 8
1001
58
NM_001201718.1
NP_001188647.1
7 - 8
1001
58
NM_001201719.1
NP_001188648.1
7 - 8
1001
58
NM_001201720.1
NP_001188649.1
7 - 8
1001
58
NM_001201721.1
NP_001188650.1
7 - 8
1001
58
NM_001201722.1
NP_001188651.1
7 - 8
1001
58
NM_001201723.1
NP_001188652.1
7 - 8
1001
58
NM_001272690.1
NP_001259619.1
8 - 9
1034
58
NM_001298403.1
NP_001285332.1
8 - 9
1187
58
NM_001298404.1
NP_001285333.1
7 - 8
998
58
NM_078647.4
NP_523371.2
8 - 9
1034
58
Back To Top
More Information
Set Membership:
Amplicon length less than or equal to 70
Drosophila melanogaster
Panther Classification:
Molecular Function -
voltage-gated ion channel
Gene Ontology Categories:
Function(s)
Process(es)
response to hypoxia
mechanosensory behavior
response to mechanical stimulus
neuronal action potential
sodium ion transmembrane transport
male courtship behavior, veined wing generated song production
regulation of postsynaptic membrane potential
membrane depolarization during action potential
voltage-gated sodium channel activity
cation channel activity
sodium channel activity
calcium ion binding
Back To Top
Related Products
TaqMan
®
Fast Advanced Master Mix
SuperScript
®
VILO
™
cDNA Synthesis Kit
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-75dc5564b9-rzq2w:80/10.239.163.191:80.
git-commit: 1b386f5412c20333c1bb415eea69346c63d8df9e
git-url: git@github.com:thermofisher/magellan-core.git
git-branch: origin/release/2.7.0-2024.04.17-1.0