Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fim | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_078661.4 | NP_523385.1 | 3 - 4 | 1961 | 69 | |
NM_167564.2 | NP_728073.1 | 4 - 5 | 1843 | 69 | ||
NM_167565.2 | NP_728074.1 | 3 - 4 | 1810 | 69 | ||
GenBank mRNA | AF053225.1 | - | 3 - 4 | 1928 | 69 | |
AY069342.1 | - | 3 - 4 | 1972 | 69 | ||
BT133379.1 | - | 3 - 4 | 1784 | 69 | ||
BT133476.1 | - | 4 - 5 | 1843 | 69 |
Set Membership: |
Amplicon length less than or equal to 70 Drosophila melanogaster |