Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: beta-Spec | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001272731.1 | NP_001259660.1 | 2 - 3 | 6407 | 56 | |
NM_001272732.1 | NP_001259661.1 | 2 - 3 | 6411 | 56 | ||
NM_078664.3 | NP_523388.1 | 2 - 3 | 6411 | 56 |
Set Membership: |
Amplicon length less than or equal to 70 Drosophila melanogaster |