Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Bx | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001272758.1 | NP_001259687.1 | 3 - 4 | 1326 | 79 | |
NM_001272759.2 | NP_001259688.2 | 4 - 5 | 1141 | 79 | ||
NM_078678.4 | NP_523402.4 | 3 - 4 | 883 | 79 | ||
NM_167625.3 | NP_728184.1 | 3 - 4 | 1326 | 79 | ||
NM_176758.3 | NP_788931.1 | 4 - 5 | 1162 | 79 | ||
GenBank mRNA | AY060902.1 | - | 3 - 4 | 1258 | 79 | |
BT004844.1 | - | 3 - 4 | 852 | 79 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Recommended Assay Drosophila melanogaster |