Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hlc | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001298588.1 | NP_001285517.1 | 2 - 3 | 910 | 65 | |
NM_078710.4 | NP_523434.2 | 2 - 3 | 910 | 65 | ||
GenBank mRNA | AY070624.1 | - | 2 - 3 | 901 | 65 |
Set Membership: |
Amplicon length less than or equal to 70 Recommended Assay Drosophila melanogaster |